Få mere at vide
N,O-Bis(trimethyl-d9-silyl)acetamide, CDN
Artikelnummer. 30322599
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322599 0.25 g
30318745 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322599 Leverandør CDN Leverandørnr. CDND10600.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,O-Bis(trimethyl-d9-silyl)acetamide
CAS 203784-65-0
Molekylær formel C8 2H18 H3 N O Si2
Mængde 0.25 g
InChI formel InChI=1 S/C8H21NOSi2/c1-8(9-11(2,3)4)10-12(5,6)7/h1-7H3/b9-8+/i2D3,3D3,4D3,5D3,6D3,7D3
SMIL [2 H]C([2 H])([2 H])[Si](O\C(=N\[Si](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])\C)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn tris(trideuteriomethyl)silyl (1 E)-N-[tris(trideuteriomethyl)silyl]ethanimidate
Molekylvægt (g/mol) 221.54
Formel vægt 221.2291 g/mol
Procent renhed 99 atom % D, min 94% Chemical Purity