Få mere at vide
N,O-Bis(trimethyl-d9-silyl)acetamide >90%, TRC
Artikelnummer. 30343089
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30343089 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343089 Leverandør Toronto Research Chemicals Leverandørnr. TRCB58850210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,O-Bis(trimethyl-d9-silyl)acetamide
CAS 203784-65-0
Molekylær formel C8 2H18 H3 N O Si2
Mængde 10 mg
Synonym Ethanimidic acid, N-[tri(methyl-d3)silyl]-, tri(methyl-d3)silyl ester (9CI), Tri(methyl-d3)silyl N-[tri(methyl-d3)silyl]ethanimidate, Trimethylsilyl-d9 N-(trimethylsilyl-d9)acetimidate, BSA-d18, N,O-Bis(trimethylsilyl-d9)acetamide, N,O-Bis(trimethylsilyl)acetamide-D18 (hexamethyldisilyl-D18)
InChI formel InChI=1S/C8H21NOSi2/c1-8(9-11(2,3)4)10-12(5,6)7/h1-7H3/b9-8+/i2D3,3D3,4D3,5D3,6D3,7D3
SMIL [2H]C([2H])([2H])[Si](O\C(=N\[Si](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])\C)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn tris(trideuteriomethyl)silyl (1E)-N-[tris(trideuteriomethyl)silyl]ethanimidate
Molekylvægt (g/mol) 221.54
Formel vægt 221.2291