Få mere at vide
N,O-Bis-(4-chlorobenzoyl)tyramine, Mikromol™
Artikelnummer. 17951506
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17951506 25 mg
17953246 100 mg
30462356 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17951506 Leverandør Mikromol Leverandørnr. MM0063.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N,O-Bis-(4-chlorobenzoyl)tyramine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,O-Bis-(4-chlorobenzoyl)tyramine
CAS 41859-56-7
Analyt- eller komponentnavne N,O-Bis-(4-chlorobenzoyl)tyramine
Forsendelsestilstand Room Temperature
Molekylær formel C22 H17 Cl2 N O3
Certificeringer/compliance ISO 17025
Synonym N,O-Bis-(4-chlorobenzoyl)tyramine
InChI formel InChI=1S/C22H17Cl2NO3/c23-18-7-3-16(4-8-18)21(26)25-14-13-15-1-11-20(12-2-15)28-22(27)17-5-9-19(24)10-6-17/h1-12H,13-14H2,(H,25,26)
SMIL Clc1ccc(cc1)C(=O)NCCc2ccc(OC(=O)c3ccc(Cl)cc3)cc2
IUPAC navn [4-[2-[(4-chlorobenzoyl)amino]ethyl]phenyl] 4-chlorobenzoate
Molekylvægt (g/mol) 414.28
Mængde 25 mg
Formel vægt 413.0585 g/mol