Få mere at vide
n-Nonylbenzene-d24, CDN
Artikelnummer. 30327378
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327378 0.25 g -
30317851 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327378 Leverandør CDN Leverandørnr. CDND67370.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Nonylbenzene-D24
CAS 1219799-13-9
Molekylær formel C15 D24
Mængde 0.25 g
Synonym n-Nonylbenzene-D24, Nonylbenzene-d24, Perdeuteriononylbenzene, 1-(Phenyl-d5)nonane-d19
InChI formel InChI=1 S/C15H24/c1-2-3-4-5-6-7-9-12-15-13-10-8-11-14-15/h8,10-11,13-14 H,2-7,9,12H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8 D,9D2,10 D,11 D,12D2,13 D,14 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecadeuteriononyl)benzene
Molekylvægt (g/mol) 228.499
Formel vægt 228.338 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity