Få mere at vide
n-Nonylbenzene-2,3,4,5,6-d5, CDN
Artikelnummer. 30323655
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323655 0.5 g 0.5g
30320831 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323655 Leverandør CDN Leverandørnr. CDND33410.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Nonylbenzene-2,3,4,5,6-D5
CAS 20216-93-7
Molekylær formel C15 D5 H19
Mængde 0.5 g
Synonym Nonane, 1-(phenyl-d5)- (8 CI), n-Nonylbenzene-2,3,4,5,6-D5, Nonylbenzene-d5, Nonylbenzene-d5 (phenyl-d5), 1-(Phenyl-d5)nonane
InChI formel InChI=1 S/C15H24/c1-2-3-4-5-6-7-9-12-15-13-10-8-11-14-15/h8,10-11,13-14 H,2-7,9,12H2,1H3/i8D,10 D,11 D,13 D,14 D
SMIL [2 H]c1c([2 H])c([2 H])c(CCCCCCCCC)c([2 H])c1[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-nonylbenzene
Molekylvægt (g/mol) 209.382
Formel vægt 209.219 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity