missing translation for 'onlineSavingsMsg'
Få mere at vide

n-Nonanoic Acid-d17, TRC

Artikelnummer. 30468008
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30468008 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468008 Leverandør Toronto Research Chemicals Leverandørnr. TRCN64938825MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale n-Nonanoic Acid-d17
CAS 130348-94-6
Molekylær formel C9 D17 H O2
Mængde 25 mg
Synonym 1-Octanecarboxylic Acid-d17, Emery 1203-d17, Grantrico-d17, NSC 62787-d17, NSC 65450-d17, NSC 65455-d17, Nonoic Acid-d17, Nonylic Acid-d17, Pelargonic Acid-d17, Thinex-d17, n-Nonoic Acid-d17, n-Nonylic Acid-d17, n-Pelargonic Acid-d17, Nonanoic Acid D17, Nonanoic-d17 acid (9CI), 1-Octanecarboxylic-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d17 acid
InChI formel InChI=1S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecadeuteriononanoic acid
Molekylvægt (g/mol) 175.34
Formel vægt 175.2374
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.