Få mere at vide
n-Nonanoic Acid-d17, TRC
Artikelnummer. 30484175
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30484175 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30484175 Leverandør Toronto Research Chemicals Leverandørnr. TRCN64938850MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Nonanoic Acid-d17
CAS 130348-94-6
Molekylær formel C9 D17 H O2
Mængde 50 mg
Synonym 1-Octanecarboxylic Acid-d17, Emery 1203-d17, Grantrico-d17, NSC 62787-d17, NSC 65450-d17, NSC 65455-d17, Nonoic Acid-d17, Nonylic Acid-d17, Pelargonic Acid-d17, Thinex-d17, n-Nonoic Acid-d17, n-Nonylic Acid-d17, n-Pelargonic Acid-d17, Nonanoic Acid D17, Nonanoic-d17 acid (9CI), 1-Octanecarboxylic-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d17 acid
InChI formel InChI=1S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecadeuteriononanoic acid
Molekylvægt (g/mol) 175.34
Formel vægt 175.2374