Learn More
n-Nonane, Sigma-Aldrich™
Product Code. 15896420
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
Unit Size:
1 L
100mL
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896420 250 mL 250mL
15869340 100 mL 100mL
15879340 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15896420 Supplier Sigma Aldrich Supplier No. 8068380250
 Restricted Item
Forventet forsendelse: 18-11-2026
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-84-2
Specifications
Melting Point -53°C
Density 0.72 g/cm3 at 20°C
Boiling Point 151°C/1013 hPa
Molecular Formula CH3(CH2)7CH3
MDL Number MFCD00009574
Quantity 250 mL
Synonym n-Nonane, n-Nonane
Molecular Weight (g/mol) 128.26
Grade Research