Få mere at vide
N-Nitrosopiperazine-d8, TRC
Artikelnummer. 30468462
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30468462 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468462 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5453522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Nitrosopiperazine-d8
CAS 1330180-56-7
Renhedsgrad noter GC
Molekylær formel C4HD8N3O
Mængde 2.5 mg
Synonym 1-Nitrosopiperazine-d8, N-Mononitrosopiperazine-d8, NSC 50269-d8, NSC 525340-d8
InChI formel InChI=1S/C4H9N3O/c8-6-7-3-1-5-2-4-7/h5H,1-4H2/i1D2,2D2,3D2,4D2
SMIL [2H]C1([2H])C([2H])([2H])NC([2H])([2H])C([2H])([2H])N1N=O
Molekylvægt (g/mol) 123.18
Formel vægt 123.1248
Procent renhed >95