Få mere at vide
N’-Nitrosopentyl-(2-picolyl)amine, TRC
Artikelnummer. 30478660
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30478660 25 mg
30470734 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30478660 Leverandør Toronto Research Chemicals Leverandørnr. TRCN54499025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 383417-48-9
Molekylær formel C11 H17 N3 O
Molekylvægt (g/mol) 207.27
IUPAC navn N-pentyl-N-(pyridin-2-ylmethyl)nitrous amide
SMIL CCCCCN(Cc1ccccn1)N=O
Tekniske data
Kemisk navn eller materiale N'-Nitrosopentyl-(2-picolyl)amine
Mængde 25 mg
InChI formel InChI=1S/C11H17N3O/c1-2-3-6-9-14(13-15)10-11-7-4-5-8-12-11/h4-5,7-8H,2-3,6,9-10H2,1H3
Formel vægt 207.1372