Få mere at vide
N’-Nitrosonornicotine N-Beta-D-Glucuronide (Mixture Of Diastereomers) X Hydrate, TRC
Artikelnummer. 30466752
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466752 1 mg
30474872 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30466752 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5350101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 350508-25-7
Molekylær formel C15 H19 N3 O7 . H2 O
Molekylvægt (g/mol) 371.34
Synonym Pyridinium, 1-β-D-glucopyranuronosyl-3-(1-nitroso-2-pyrrolidinyl)-, inner salt, stereoisomer (9CI), N'-Nitrosonornicotine N-β-D-Glucuronide
IUPAC navn (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[3-[(2S)-1-nitrosopyrrolidin-2-yl]pyridin-1-ium-1-yl]oxane-2-carboxylate;hydrate
SMIL O.O[C@H]1[C@H](O)[C@H](O[C@H]([C@@H]1O)[n+]2cccc(c2)[C@@H]3CCCN3N=O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale N'-Nitrosonornicotine N-beta-D-Glucuronide hydrate
Mængde 1 mg
InChI formel InChI=1S/C15H19N3O7.H2O/c19-10-11(20)13(15(22)23)25-14(12(10)21)17-5-1-3-8(7-17)9-4-2-6-18(9)16-24;/h1,3,5,7,9-14,19-21H,2,4,6H2;1H2/t9-,10-,11-,12+,13-,14+;/m0./s1
Formel vægt 371.1329