missing translation for 'onlineSavingsMsg'
Få mere at vide

N' 'Nitrosonorfloxacin, TRC

Product Code. 30481057
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30481057 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30481057 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCN13742010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Nitrosonorfloxacin
CAS 74011-53-3
Molekylær formel C16 H17 F N4 O4
Mængde 10 mg
Synonym 1-Ethyl-6-fluoro-1,4-dihydro-7-(4-nitroso-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid
InChI formel InChI=1S/C16H17FN4O4/c1-2-19-9-11(16(23)24)15(22)10-7-12(17)14(8-13(10)19)20-3-5-21(18-25)6-4-20/h7-9H,2-6H2,1H3,(H,23,24)
SMIL CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCN(CC3)N=O
IUPAC navn 1-ethyl-6-fluoro-7-(4-nitrosopiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
Molekylvægt (g/mol) 348.33
Formel vægt 348.1234
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.