Få mere at vide
N-Nitrosomorpholine-d8, CDN
Artikelnummer. 30322036
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322036 0.1 g
30316955 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322036 Leverandør CDN Leverandørnr. CDND40910.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nitrosomorpholine-d8
CAS 1219805-76-1
Molekylær formel C4 2H8 N2 O2
Mængde 0.1 g
Synonym 2,2,3,3,5,5,6,6-Octadeutero-N-nitrosomorpholine, 4-Nitrosomorpholine-2,2,3,3,5,5,6,6-d8, 4-Nitrosomorpholine D8, N-Nitrosomorpholine-2,2,3,3,5,5,6,6-d8, N-Nitrosomorpholine D8
InChI formel InChI=1 S/C4H8N2O2/c7-5-6-1-3-8-4-2-6/h1-4H2/i1D2,2D2,3D2,4D2
SMIL [2 H]C1([2 H])OC([2 H])([2 H])C([2 H])([2 H])N(N=O)C1([2 H])[2 H]
IUPAC navn 2,2,3,3,5,5,6,6-octadeuterio-4-nitrosomorpholine
Molekylvægt (g/mol) 124.17
Formel vægt 124.1088 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity