Få mere at vide
N-Nitrosoguvacine, TRC
Artikelnummer. 17923865
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17923865 5 mg
17925095 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17923865 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5276155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 55557-01-2
Molekylær formel C6H8N2O3
Molekylvægt (g/mol) 156.14
Synonym Nitrosoguvacine, 1,2,5,6-Tetrahydro-1-nitroso-3-pyridinecarboxylic Acid;
IUPAC navn 1-nitroso-3,6-dihydro-2H-pyridine-5-carboxylic acid
SMIL O=C(C1=CCCN(N=O)C1)O
Tekniske data
Kemisk navn eller materiale N-Nitrosoguvacine
Mængde 5 mg
InChI formel InChI=1S/C6H8N2O3/c9-6(10)5-2-1-3-8(4-5)7-11/h2H,1,3-4H2,(H,9,10)
Formel vægt 156.0535