Få mere at vide
N-Nitrosodimethylamine-d6 (1.0 mg/mL in Ethanol), TRC
Artikelnummer. 30471686
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30471686 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471686 Leverandør Toronto Research Chemicals Leverandørnr. TRCKIT91021X1ML
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Nitrosodi-N-methylamine-d6
CAS 17829-05-9
Molekylær formel C2 D6 N2 O
Mængde 1 mL
Synonym Di(methyl-d3)amine, N-nitroso- (8CI), Dimethylnitrosamine-d6, N-Nitrosodimethylamine-d6, N-Nitroso-dimethylamine D6, NDMA-d6, Di(methyl-d3)amine, N-nitroso- (8CI), Dimethylnitrosamine-d6, N-Nitrosodimethylamine-d6
InChI formel InChI=1S/C2H6N2O/c1-4(2)3-5/h1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])N(N=O)C([2H])([2H])[2H]
IUPAC navn N,N-bis(trideuteriomethyl)nitrous amide
Molekylvægt (g/mol) 80.12
Formel vægt 80.0857
Koncentration eller sammensætning (efter analyt eller komponenter) N-Nitrosodi-N-methylamine-d6 1.0 mg/mL