Få mere at vide
N-Nitrosodiisobutylamine-d4, TRC
Artikelnummer. 30472309
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30472309 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472309 Leverandør Toronto Research Chemicals Leverandørnr. TRCN52561225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Nitrosodiisobutylamine-d4
CAS 1794897-96-3
Renhedsgrad noter GC
Molekylær formel C8H14D4N2O
Mængde 25 mg
Synonym 2-Methyl-N-(2-methylpropyl)-N-nitroso-1-propanamine-d4, 2,2'-Dimethyl-N-nitrosodipropylamine-d4, Diisobutylnitrosamine-d4, Diisobutylnitrosoamine-d4, N,N-Diisobutylnitrosamine-d4, N-Nitroso-di-iso-butylamine-d4, NSC 134-d4
InChI formel InChI=1S/C8H18N2O/c1-7(2)5-10(9-11)6-8(3)4/h7-8H,5-6H2,1-4H3/i5D2,6D2
SMIL [2H]C(C(C)C)([2H])N(N=O)C([2H])([2H])C(C)C
Molekylvægt (g/mol) 162.27
Formel vægt 162.167
Procent renhed >95