Få mere at vide
N-Nitrosodiethylamine-d4 + Nitrosobis(2-hydroxyethyl)amine-d8 (Mixture) (1mg/mL in Methanol), TRC
Artikelnummer. 30483971
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30483971 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483971 Leverandør Toronto Research Chemicals Leverandørnr. TRCKIT65281X1ML
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nitrosobis(2-hydroxyethyl)amine-d8∣N-Nitroso-diethylamine (dimethylene-D4)
CAS 1173019-53-8, 1346603-41-5
Opløsningsmiddel eller Matrix Methanol
Mængde 1 mL
Koncentration eller sammensætning (efter analyt eller komponenter) Nitrosobis(2-hydroxyethyl)amine-d8 1mg/mL∣N-Nitroso-diethylamine (dimethylene-D4) 1mg/mL