Få mere at vide
N-Nitrosodibenzylamine-d4, TRC
Artikelnummer. 30482054
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30482054 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482054 Leverandør Toronto Research Chemicals Leverandørnr. TRCN52545350MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Nitrosodibenzylamine D4 (Dimethylen D4)
CAS 20002-23-7
Renhedsgrad noter HPLC
Molekylær formel C14 D4 H10 N2 O
Mængde 50 mg
Synonym N-Nitrosodibenzylamine D4 (Dimethylen D4)
InChI formel InChI=1S/C14H14N2O/c17-15-16(11-13-7-3-1-4-8-13)12-14-9-5-2-6-10-14/h1-10H,11-12H2/i11D2,12D2
SMIL [2H]C([2H])(N(N=O)C([2H])([2H])c1ccccc1)c2ccccc2
IUPAC navn N,N-bis[dideuterio(phenyl)methyl]nitrous amide
Molekylvægt (g/mol) 230.298
Formel vægt 230.136
Procent renhed >95