Få mere at vide
N-Nitrosodesmethylchlorpromazine, Mikromol
Beskrivelse
N-Nitrosodesmethylchlorpromazine, Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Nitrosodesmethylchlorpromazine |
| CAS | 55855-44-2 |
| Analyt- eller komponentnavne | N-Nitrosodesmethylchlorpromazine |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H16 Cl N3 O S |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 10H-Phenothiazine-10-propanamine, 2-chloro-N-methyl-N-nitroso-, 2-Chloro-N-methyl-N-nitroso-10H-phenothiazine-10-propanamine, N-Nitrosodesmethylchlorpromazine, [3-(2-Chloro-phenothiazin-10-yl)-propyl]-methyl-nitroso-amine |
| InChI formel | InChI=1S/C16H16ClN3OS/c1-19(18-21)9-4-10-20-13-5-2-3-6-15(13)22-16-8-7-12(17)11-14(16)20/h2-3,5-8,11H,4,9-10H2,1H3 |
| Løsningstype | Neat |
| SMIL | CN(CCCN1c2ccccc2Sc3ccc(Cl)cc13)N=O |
| Vis mere |