Få mere at vide
N-Nitrosobis(2-hydroxyethyl)-d8-amine, CDN
Artikelnummer. 30319351
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319351 0.1 g
30309659 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319351 Leverandør CDN Leverandørnr. CDND59970.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nitrosobis(2-hydroxyethyl)amine-d8
CAS 1173019-53-8
Molekylær formel C4D8H2N2O3
Mængde 0.1 g
Synonym 2,2’-(Nitrosoimino)bisethanol-d8, Diethanolnitrosamine-d8, N,N-Diethanolnitrosamine-d8, N-Nitrosodiethanolamine-d8, NDELA-d8;
InChI formel InChI=1 S/C4H10N2O3/c7-3-1-6(5-9)2-4-8/h7-8 H,1-4H2/i1D2,2D2,3D2,4D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])N(N=O)C([2 H])([2 H])C([2 H])([2 H])O
IUPAC navn N,N-bis(1,1,2,2-tetradeuterio-2-hydroxy-ethyl)nitrous amide
Molekylvægt (g/mol) 142.18
Formel vægt 142.1194 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity