missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Nitroso Safinamide, TRC

Artikelnummer. 30410754
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30410754 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410754 Supplier Toronto Research Chemicals Supplier No. TRCN703665100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Nitroso Safinamide
CAS 2657645-00-4
Molekylær formel C17H18FN3O3
Mængde 100 mg
Synonym Propanamide','2-[[[4-[(3-fluorophenyl)methoxy]phenyl]methyl]nitrosoamino]-','(2S)- (ACI), (2S)-2-[[[4-[(3-Fluorophenyl)methoxy]phenyl]methyl]nitrosoamino]propanamide (ACI), N-Nitroso Safinamide
InChI formel InChI=1S/C17H18FN3O3/c1-12(17(19)22)21(20-23)10-13-5-7-16(8-6-13)24-11-14-3-2-4-15(18)9-14/h2-9,12H,10-11H2,1H3,(H2,19,22)/t12-/m0/s1
SMIL C[C@H](N(CC1=CC=C(OCC2=CC=CC(F)=C2)C=C1)N=O)C(N)=O
IUPAC navn (S)-2-((4-((3-fluorobenzyl)oxy)benzyl)(nitroso)amino)propanamide
Molekylvægt (g/mol) 331.35
Formel vægt 331.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.