Få mere at vide
N-Nitroso Nipecotic Acid-d4 (Major), TRC
Artikelnummer. 30476786
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30476786 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476786 Leverandør Toronto Research Chemicals Leverandørnr. TRCN53200210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Nitroso Nipecotic Acid-d4 (Major)
CAS 1329834-25-4
Molekylær formel C6H6D4N2O3
Mængde 10 mg
Synonym 1-Nitroso-3-piperidinecarboxylic Acid-d4, 1-Nitrosonipecotic Acid-d4
InChI formel InChI=1S/C6H10N2O3/c9-6(10)5-2-1-3-8(4-5)7-11/h5H,1-4H2,(H,9,10)/i3D2,4D2
SMIL [2H]C1([2H])CCC(C(=O)O)C([2H])([2H])N1N=O
IUPAC navn 2,2,6,6-tetradeuterio-1-nitrosopiperidine-3-carboxylic acid
Molekylvægt (g/mol) 162.18
Formel vægt 162.0942