Få mere at vide
N-Nitroso-N,N-di(3,5,5-trimethylhexyl)amine-d4, TRC
Artikelnummer. 30466607
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30466607 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466607 Leverandør Toronto Research Chemicals Leverandørnr. TRCN52580225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Nitroso-N,N-di(3,5,5-trimethylhexyl)amine-d4
CAS 1346603-10-8
Molekylær formel C18H34D4N2O
Mængde 25 mg
Synonym 3,5,5-Trimethyl-N-nitroso-N-(3,5,5-trimethylhexyl)-1-hexanamine-d4, Bis(3,5,5-trimethylhexyl)-N-nitrosoamine-d4
InChI formel InChI=1S/C18H38N2O/c1-15(13-17(3,4)5)9-11-20(19-21)12-10-16(2)14-18(6,7)8/h15-16H,9-14H2,1-8H3/i11D2,12D2
SMIL [2H]C(N(N=O)C([2H])([2H])CC(C)CC(C)(C)C)([2H])CC(C)CC(C)(C)C
Molekylvægt (g/mol) 302.53
Formel vægt 302.3235