Få mere at vide
N-Nitroso-N-n-butyl-N-methylamine, Mikromol™
Beskrivelse
N-Nitroso-N-n-butyl-N-methylamine
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Butyl-N-methylnitrosamine |
| CAS | 7068-83-9 |
| Analyt- eller komponentnavne | N-Butyl-N-methylnitrosamine |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C5 H12 N2 O |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1-Butanamine, N-methyl-N-nitroso-, Butylamine, N-methyl-N-nitroso- (6CI,7CI,8CI), N-Methyl-N-nitroso-1-butanamine, Methylbutylnitrosamine, N-Butyl-N-methylnitrosamine, N-Methyl-N-butylnitrosamine, N-Methyl-N-nitrosobutylamine, N-Nitroso-N-methylbutylamine, N-Nitrosobutylmethylamine, N-Nitrosomethylbutylamine, Nitrosomethylbutylamine, n-Butylmethylnitrosamine, N-Nitroso-N-n-butyl-N-methylamine, NBMA |
| InChI formel | InChI=1S/C5H12N2O/c1-3-4-5-7(2)6-8/h3-5H2,1-2H3 |
| SMIL | CCCCN(C)N=O |
| IUPAC navn | N-butyl-N-methylnitrous amide |
| Vis mere |