Få mere at vide
N-Nitroso-N-methylbenzylamine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Nitroso-N-methylbenzylamine |
| CAS | 937-40-6 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C8 H10 N2 O |
| Mængde | 1 g |
| Synonym | Benzenemethanamine, N-methyl-N-nitroso-, Benzylamine, N-methyl-N-nitroso- (6CI,7CI,8CI), N-Methyl-N-nitrosobenzenemethanamine, Benzyl(methyl)nitrosamine, Methyl(benzyl)nitrosamine, N-Methyl-N-benzylnitrosamine, N-Methyl-N-nitrosobenzylamine, N-Nitroso(benzyl)methylamine, N-Nitroso(methyl)benzylamine, N-Nitroso-N-methylbenzenemethanamine, N-Nitroso-N-methylbenzylamine, NMBA |
| InChI formel | InChI=1S/C8H10N2O/c1-10(9-11)7-8-5-3-2-4-6-8/h2-6H,7H2,1H3 |
| SMIL | CN(Cc1ccccc1)N=O |
| IUPAC navn | N-benzyl-N-methylnitrous amide |
| Molekylvægt (g/mol) | 150.1778 |
| Vis mere |