Få mere at vide
N-Nitroso-N-methyl-DL-phenylalanine, TRC
Artikelnummer. 30471953
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471953 10 mg
30465560 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30471953 Leverandør Toronto Research Chemicals Leverandørnr. TRCN52995010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 41867-08-7
Molekylær formel C10 H12 N2 O3
Molekylvægt (g/mol) 208.21
IUPAC navn 2-[methyl(nitroso)amino]-3-phenylpropanoic acid
SMIL CN(N=O)C(Cc1ccccc1)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Nitroso-N-methyl-DL-phenylalanine
InChI formel InChI=1S/C10H12N2O3/c1-12(11-15)9(10(13)14)7-8-5-3-2-4-6-8/h2-6,9H,7H2,1H3,(H,13,14)
Mængde 10 mg
Formel vægt 208.0848