Få mere at vide
N-Nitroso-N-methyl-4-aminobutyric acid D3 (N-methyl D3), Dr. Ehrenstorfer
Artikelnummer. 30325544
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30325544 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325544 Leverandør Dr. Ehrenstorfer Leverandørnr. DREC15605410
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Nitroso-N-(methyl-d3)-4-aminobutyric Acid
CAS 1184996-41-5
Molekylær formel C5 2H3 H9 N2 O
Mængde 10 mg
InChI formel InChI=1S/C5H12N2O/c1-3-4-5-7(2)6-8/h3-5H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])N(CCCC)N=O
IUPAC navn N-butyl-N-(trideuteriomethyl)nitrous amide
Molekylvægt (g/mol) 119.18
Formel vægt 119.1138 g/mol