missing translation for 'onlineSavingsMsg'
Learn More

N-nitroso-N-methyl-3-aminopropionsyre, methylester, TRC

Product Code. 30473439
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30473439 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473439 Supplier Toronto Research Chemicals Supplier No. TRCN52925025MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Nitroso-N-methyl-3-aminopropionic Acid','Methyl Ester
CAS 383417-47-8
Molekylær formel C5 H10 N2 O3
Mængde 25 mg
Synonym Propanoic acid','3-(methylnitrosoamino)-','methyl ester (ACI), Methyl 3-(methylnitrosoamino)propanoate (ACI), β-Alanine','N-methyl-N-nitroso-','methyl ester (9CI), 3-(Methylnitrosoamino)propanoic acid methyl ester, N-Methyl-N-nitroso-β-alanine methyl ester, N-Nitroso-N-methyl-3-aminopropionic acid methyl ester
InChI formel InChI=1S/C5H10N2O3/c1-7(6-9)4-3-5(8)10-2/h3-4H2,1-2H3
SMIL COC(=O)CCN(C)N=O
IUPAC navn Methyl 3-[methyl(nitroso)amino]propanoat
Molekylvægt (g/mol) 146.14
Formel vægt 146.0691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.