Få mere at vide
N-Nitroso-N-methyl-3-aminopropionic Acid-d3, TRC
Artikelnummer. 30482051
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30482051 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482051 Leverandør Toronto Research Chemicals Leverandørnr. TRCN52920250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Nitroso-N-(methyl-d3)-3-aminopropionic Acid
CAS 1215691-18-1
Molekylær formel C4 2H3 H5 N2 O3
Mængde 50 mg
Synonym 3-(Nitroso-(trideuteriomethyl)amino)propanoic acid, 3-(Methyl-d3-nitrosoamino)propanoic acid, 3-[N-Nitroso-N-(methyl-d3)amino]propionic acid, NMPA-d3, N-Nitroso-N-(methyl-d3)-3-aminopropionic acid, N-(Methyl-d3)-N-nitroso-β-alanine, 3-(N-Nitroso-N-methylamino)propanoic acid D3 (N-methyl-D3)
InChI formel InChI=1S/C4H8N2O3/c1-6(5-9)3-2-4(7)8/h2-3H2,1H3,(H,7,8)/i1D3
SMIL [2H]C([2H])([2H])N(CCC(=O)O)N=O
IUPAC navn 3-[nitroso(trideuteriomethyl)amino]propanoic acid
Molekylvægt (g/mol) 135.14
Formel vægt 135.0723