missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Nitroso Lisinopril, TRC

Artikelnummer. 30469421
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469421 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30469421 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4689935MG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Nitroso Lisinopril
CAS 519175-80-5
Renhedsgrad noter HPLC
Molekylær formel C21 H30 N4 O6
Mængde 5 mg
Synonym N-Nitrosolisinopril, L-Proline, N2-[(1S)-1-carboxy-3-phenylpropyl]-N2-nitroso-L-lysyl- (9CI)
InChI formel InChI=1S/C21H30N4O6/c22-13-5-4-9-16(19(26)24-14-6-10-17(24)20(27)28)25(23-31)18(21(29)30)12-11-15-7-2-1-3-8-15/h1-3,7-8,16-18H,4-6,9-14,22H2,(H,27,28)(H,29,30)/t16-,17-,18-/m0/s1
SMIL NCCCC[C@H](N(N=O)[C@@H](CCc1ccccc1)C(=O)O)C(=O)N2CCC[C@H]2C(=O)O
IUPAC navn (2S)-1-[(2S)-6-amino-2-[[(1S)-1-carboxy-3-phenylpropyl]-nitrosoamino]hexanoyl]pyrrolidine-2-carboxylic acid
Molekylvægt (g/mol) 434.486
Formel vægt 434.217
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.