Få mere at vide
N-Nitroso Guvacoline-d4 (major), TRC
Artikelnummer. 30474222
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30474222 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474222 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5276271MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Nitroso Guvacoline-d4 (major)
CAS 1330277-21-8
Molekylær formel C7H6D4N2O3
Mængde 1 mg
Synonym 1,2,5,6-Tetrahydro-1-nitroso-3-pyridinecarboxylic Acid-d4 Methyl Ester, Nitrosoguvacoline-d4
InChI formel InChI=1S/C7H10N2O3/c1-12-7(10)6-3-2-4-9(5-6)8-11/h3H,2,4-5H2,1H3/i2D2,5D2
SMIL [2H]C1([2H])CN(N=O)C([2H])([2H])C(=C1)C(=O)OC
IUPAC navn methyl 3,3,6,6-tetradeuterio-1-nitroso-2H-pyridine-5-carboxylate
Molekylvægt (g/mol) 174.19
Formel vægt 174.0942