Få mere at vide
N-Nitroso-diphenylamine D10, Dr. Ehrenstorfer
Artikelnummer. 30321019
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30321019 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321019 Leverandør Dr. Ehrenstorfer Leverandørnr. DRECA15604510
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Nitrosodiphenylamine-d10
CAS 42952-91-0
Molekylær formel C12 2H10 N2 O
Mængde 10 mg
Synonym Benzen-d5-amine, N-nitroso-N-(phenyl-d5)- (9CI), N-Nitroso-diphenylamine D10, N-Nitroso-bis(pentadeuteriophenyl)amine
InChI formel InChI=1S/C12H10N2O/c15-13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c([2H])c(N(N=O)c2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H]
IUPAC navn N,N-bis(2,3,4,5,6-pentadeuteriophenyl)nitrous amide
Molekylvægt (g/mol) 208.28
Formel vægt 208.1421 g/mol