Få mere at vide
N-Nitroso(2-hydroxyethyl)glycine, TRC
Artikelnummer. 30137044
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137044 100 mg
30011449 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137044 Leverandør Toronto Research Chemicals Leverandørnr. TRCN528400100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 80556-89-4
Molekylær formel C4H8N2O4
Molekylvægt (g/mol) 148.12
Synonym Acetic acid, 2-[(2-hydroxyethyl)nitrosoamino]- (ACI), 2-[(2-Hydroxyethyl)nitrosoamino]acetic acid (ACI), Glycine, N-(2-hydroxyethyl)-N-nitroso- (7CI, 9CI), N-(2-Hydroxyethyl)-N-carboxymethylnitrosamine, N-Nitroso(2-hydroxyethyl)glycine, N-(2-Hydroxyethyl)-N-nitrosoglycine
IUPAC navn 2-[2-hydroxyethyl(nitroso)amino]acetic acid
SMIL OCCN(CC(=O)O)N=O
Tekniske data
Kemisk navn eller materiale N-Nitroso(2-hydroxyethyl)glycine
InChI formel InChI=1S/C4H8N2O4/c7-2-1-6(5-10)3-4(8)9/h7H,1-3H2,(H,8,9)
Mængde 100 mg
Formel vægt 148.0484