missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Nitroso-1-(2-methoxyphenyl)piperazine, TRC

Artikelnummer. 30427426
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427426 250 mg
30424798 500 mg
30414259 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30427426 Leverandør Toronto Research Chemicals Leverandørnr. TRCN754470250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2219339-64-5
Molekylær formel C11H15N3O2
Molekylvægt (g/mol) 221.26
Synonym 1-(2-Methoxyphenyl)-4-nitroso-piperazine
IUPAC navn 1-(2-methoxyphenyl)-4-nitrosopiperazine
SMIL COc1ccccc1N2CCN(CC2)N=O

Specifications

Kemisk navn eller materiale N-Nitroso-1-(2-methoxyphenyl)piperazine
Mængde 250 mg
InChI formel InChI=1S/C11H15N3O2/c1-16-11-5-3-2-4-10(11)13-6-8-14(12-15)9-7-13/h2-5H,6-9H2,1H3
Formel vægt 221.1164
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.