Få mere at vide
N-Nitroso-1-(2-methoxyphenyl)piperazine, TRC
Artikelnummer. 30424798
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424798 500 mg
30427426 250 mg
30414259 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30424798 Leverandør Toronto Research Chemicals Leverandørnr. TRCN754470500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2219339-64-5
Molekylær formel C11H15N3O2
Molekylvægt (g/mol) 221.26
Synonym 1-(2-Methoxyphenyl)-4-nitroso-piperazine
IUPAC navn 1-(2-methoxyphenyl)-4-nitrosopiperazine
SMIL COc1ccccc1N2CCN(CC2)N=O
Tekniske data
Kemisk navn eller materiale N-Nitroso-1-(2-methoxyphenyl)piperazine
Mængde 500 mg
InChI formel InChI=1S/C11H15N3O2/c1-16-11-5-3-2-4-10(11)13-6-8-14(12-15)9-7-13/h2-5H,6-9H2,1H3
Formel vægt 221.1164