missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Nitro-Alpha-[2-(Methylamino)ethyl]benzyl Alcohol, TRC

Artikelnummer. 30179261
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179261 250 mg
30179309 100 mg
30178923 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30179261 Leverandør Toronto Research Chemicals Leverandørnr. TRCN501015250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2659259-65-9
Molekylær formel C10H14N2O2
Molekylvægt (g/mol) 194.23
Synonym N-(3-Hydroxy-3-phenylpropyl)-N-methylnitrous Amide
IUPAC navn N-(3-hydroxy-3-phenylpropyl)-N-methylnitrous amide
SMIL CN(CCC(C1=CC=CC=C1)O)N=O

Specifications

Kemisk navn eller materiale N-Nitro-alpha-[2-(Methylamino)ethyl]benzyl Alcohol
Mængde 250 mg
InChI formel InChI=1S/C10H14N2O2/c1-12(11-14)8-7-10(13)9-5-3-2-4-6-9/h2-6,10,13H,7-8H2,1H3
Formel vægt 194.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.