missing translation for 'onlineSavingsMsg'
Få mere at vide

N,n,n-trimethyl-1,3-propanediamine, 98%, BLD Pharm™

Product Code. 30988802
Encompass_Preferred
Change view
Click to view available options
Mængde:
1 g
5 g
25 g
Unit Size:
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde Unit Size
30988802 25 g 25g
30972458 1 g 1g
30971935 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30988802 Supplier BLD Pharm Supplier No. BD14180125g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale N,n,n-trimethyl-1,3-propanediamine
CAS 4543-96-8
Molekylær formel C6H16N2
MDL nummer MFCD00014879
Mængde 25 g
SMIL CN(C)CCCNC
Molekylvægt (g/mol) 116.20
Procent renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.