missing translation for 'onlineSavingsMsg'
Få mere at vide

N,N,N-Tri(benzyloxycarbonyl) Deoxypyridinoline Tri-(tert-butyl) Ester, TRC

Product Code. 30457130
Change view
Click to view available options
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30457130 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457130 Supplier Toronto Research Chemicals Supplier No. TRCT7715501MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N,N,N-Tri(benzyloxycarbonyl) Deoxypyridinoline Tri-(tert-butyl) Ester
CAS 224629-59-8
Molekylær formel C54H70N4O13
Mængde 1 mg
Synonym 3-[(3S)-4-(1,1-Dimethylethoxy)-4-oxo-3-[[(phenylmethoxy)carbonyl]amino]butyl]-1-[(5S)-6-(1,1-dimethylethoxy)-6-oxo-5-[[(phenylmethoxy)carbonyl]amino]hexyl]-4-[(2S)-3-(1,1-dimethylethoxy)-3-oxo-2-[[(phenylmethoxy)carbonyl]amino]propyl]-5-hydroxy-pyridinium Inner Salt
InChI formel InChI=1S/C54H70N4O13/c1-52(2,3)69-46(60)42(55-49(63)66-34-37-21-13-10-14-22-37)27-19-20-30-58-32-40(28-29-43(47(61)70-53(4,5)6)56-50(64)67-35-38-23-15-11-16-24-38)41(45(59)33-58)31-44(48(62)71-54(7,8)9)57-51(65)68-36-39-25-17-12-18-26-39/h10-18,21-26,32-33,42-44H,19-20,27-31,34-36H2,1-9H3,(H3-,55,56,57,59,63,64,65)/t42-,43-,44-/m0/s1
SMIL CC(C)(C)OC(=O)[C@H](CCCC[n+]1cc([O-])c(C[C@H](NC(=O)OCc2ccccc2)C(=O)OC(C)(C)C)c(CC[C@H](NC(=O)OCc3ccccc3)C(=O)OC(C)(C)C)c1)NC(=O)OCc4ccccc4
IUPAC navn 5-[(3S)-4-[(2-methylpropan-2-yl)oxy]-4-oxo-3-(phenylmethoxycarbonylamino)butyl]-1-[(5S)-6-[(2-methylpropan-2-yl)oxy]-6-oxo-5-(phenylmethoxycarbonylamino)hexyl]-4-[(2S)-3-[(2-methylpropan-2-yl)oxy]-3-oxo-2-(phenylmethoxycarbonylamino)propyl]pyridin-1-ium-3-olat
Molekylvægt (g/mol) 983.15
Formel vægt 982.4939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.