Få mere at vide
N,N,N',N'-Tetramethyl-D-tartaramide, TRC
Artikelnummer. 30449532
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30449532 10 g
30456074 1 g
30450569 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30449532 Leverandør Toronto Research Chemicals Leverandørnr. TRCT30535010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 63126-52-3
Molekylær formel C8H16N2O4
Molekylvægt (g/mol) 204.22
Synonym (2S,3S)-2,3-Dihydroxy-N,N,N',N'-tetramethyl-butanediamide, [S-(R*,R*)]-2,3-Dihydroxy-N,N,N',N'-tetramethyl-butanediamide, (-)-D-Tartaric Acid Bis(N,N-dimethylamide), (-)-Tetramethyl-(D)-tartaric Acid Diamide, N,N,N',N'-Tetramethyl-(S,S)-tartaramide
IUPAC navn (2S,3S)-2,3-dihydroxy-N,N,N',N'-tetramethylbutanediamide
SMIL CN(C)C(=O)[C@@H](O)[C@H](O)C(=O)N(C)C
Tekniske data
Kemisk navn eller materiale N,N,N',N'-Tetramethyl-D-tartaramide
Mængde 10 g
InChI formel InChI=1S/C8H16N2O4/c1-9(2)7(13)5(11)6(12)8(14)10(3)4/h5-6,11-12H,1-4H3/t5-,6-/m0/s1
Formel vægt 204.111