Få mere at vide
N-[[N-Methyl-N-[(2-isopropyl-1,1,1,3,3,3-d6]-4-thiazolyl)methyl)amino]carbonyl-L-valine Carboxylic Acid, TRC
Artikelnummer. 30470227
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30470227 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470227 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3148525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-((N-Methyl-N-((2-isopropyl-1,1,1,3,3,3-d6)-4-thiazolyl)methyl)amino)carbonyl-L-valine Carboxylic Acid
CAS 1331906-73-0
Molekylær formel C14H17D6N3O3S
Mængde 5 mg
Synonym N-[[Methyl[[2-(1-methylethyl)-4-thiazolyl]methyl]amino]carbonyl]-L-valine-d6
InChI formel InChI=1S/C14H23N3O3S/c1-8(2)11(13(18)19)16-14(20)17(5)6-10-7-21-12(15-10)9(3)4/h7-9,11H,6H2,1-5H3,(H,16,20)(H,18,19)/t11-/m0/s1/i3D3,4D3
SMIL [2H]C([2H])([2H])C(c1nc(CN(C)C(=O)N[C@@H](C(C)C)C(=O)O)cs1)C([2H])([2H])[2H]
IUPAC navn (2S)-2-[[[2-(1,1,1,3,3,3-hexadeuteriopropan-2-yl)-1,3-thiazol-4-yl]methyl-methylcarbamoyl]amino]-3-methylbutanoic acid
Molekylvægt (g/mol) 319.45
Formel vægt 319.1837