Få mere at vide
N,N’-Ethylenebis(iodoacetamide), TRC
Artikelnummer. 30012757
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012757 10 mg
30010363 50 mg
30009654 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012757 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91750010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7250-43-3
Molekylær formel C6 H10 I2 N2 O2
Molekylvægt (g/mol) 395.96
IUPAC navn 2-iodo-N-[2-[(2-iodoacetyl)amino]ethyl]acetamide
SMIL ICC(=O)NCCNC(=O)CI
Tekniske data
Kemisk navn eller materiale N,N'-Ethylenebis(iodoacetamide)
Mængde 10 mg
InChI formel InChI=1S/C6H10I2N2O2/c7-3-5(11)9-1-2-10-6(12)4-8/h1-4H2,(H,9,11)(H,10,12)
Formel vægt 395.8832