Learn More
N,N′-Diphenylbenzidine, Sigma-Aldrich™
Product Code. 15841011
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity Unit Size
15841011 10 g 10g
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15841011 Supplier Sigma Aldrich Supplier No. 8205300010
 Restricted Item
Forventet forsendelse: 24-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 531-91-9
Melting Point 245°C to 250°C
Molecular Formula 4-(C6H5NH)C6H4C6H4-4-(NHC6H5)
MDL Number MFCD00003016
Quantity 10 g
Synonym N,N′-Diphenylbenzidine, N,N′-Diphenyl-4,4′-diaminobiphenyl, N,N′-Diphenyl-[1,1′-biphenyl]-4,4′-diamine, N,N′-Diphenylbenzidine
Molecular Weight (g/mol) 336.43
Grade Research