Få mere at vide
N,N-Dimethylsulfamide-d6, TRC
Artikelnummer. 30371009
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30371009 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371009 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4792822.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethylsulfamide-d6
CAS 1189689-95-9
Molekylær formel C2 D6 H2 N2 O2 S
Mængde 2.5 mg
Synonym N,N-Di(methyl-d3)sulfamide, N,N-Dimethylsulfamide-d6, N,N-Dimethylsulfamide-D6 (N,N-dimethyl-D6), DMSA-d6, Dimethylsulfamide-d6, 1,1-Dimethyl-d6-sulfamide, Trideuterio-[sulfamoyl(trideuteriomethyl)amino]methane
InChI formel InChI=1S/C2H8N2O2S/c1-4(2)7(3,5)6/h1-2H3,(H2,3,5,6)/i1D3,2D3
SMIL [2H]C([2H])([2H])N(C([2H])([2H])[2H])S(=O)(=O)N
IUPAC navn trideuterio-[sulfamoyl(trideuteriomethyl)amino]methane
Molekylvægt (g/mol) 130.199
Formel vægt 130.068