Få mere at vide
N,N'-Dimethylpiperazine, TRC
Artikelnummer. 30375908
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30375908 500 mg
30378142 1 g
30372818 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30375908 Leverandør Toronto Research Chemicals Leverandørnr. TRCD481298500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 106-58-1
Molekylær formel C6 H14 N2
Molekylvægt (g/mol) 114.19
Synonym N,N'-Dimethylpiperazine, Piperazine, 1,4-dimethyl-, 1,4-Dimethylpiperazine, Addocat 117, Jeffcat DMP, Lupetazine, N,N'-Dimethylpiperazine, NSC 41177, RCA 117, Texacat DMP, AH 1243X
IUPAC navn 1,4-dimethylpiperazine
SMIL CN1CCN(C)CC1
Tekniske data
Kemisk navn eller materiale N,N'-Dimethylpiperazine
Mængde 500 mg
InChI formel InChI=1S/C6H14N2/c1-7-3-5-8(2)6-4-7/h3-6H2,1-2H3
Formel vægt 114.1157