Få mere at vide
N,N-Dimethylmethanamine Vonoprazan, TRC
Artikelnummer. 30134061
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134061 50 mg
30007190 100 mg
17939094 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134061 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47704850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1885094-62-1
Molekylær formel C18 H18 F N3 O2 S
Molekylvægt (g/mol) 359.42
Synonym 5-(2-Fluorophenyl)-N,N-dimethyl-1-(3-pyridinylsulfonyl)-1H-pyrrole-3-methanamine
IUPAC navn 1-[5-(2-fluorophenyl)-1-(3-pyridylsulfonyl)pyrrol-3-yl]-N,N-dimethyl-methanamine
SMIL CN(C)Cc1cc(c2ccccc2F)n(c1)S(=O)(=O)c3cccnc3
Tekniske data
Kemisk navn eller materiale N,N-Dimethylmethanamine Vonoprazan
Mængde 50 mg
InChI formel InChI=1S/C18H18FN3O2S/c1-21(2)12-14-10-18(16-7-3-4-8-17(16)19)22(13-14)25(23,24)15-6-5-9-20-11-15/h3-11,13H,12H2,1-2H3
Formel vægt 359.1104