Få mere at vide
N,N-Dimethylmethanamine Oxide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N,N-Dimethylmethanamine Oxide |
| CAS | 1184-78-7 |
| Molekylær formel | C3H9NO |
| Mængde | 500 mg |
| Synonym | Trimethylamine, N-oxide (6CI, 8CI), N,N-Dimethylmethanamine N-oxide, NSC 408426, TMAO, Trimethylamine oxide, Triox |
| InChI formel | InChI=1S/C3H9NO/c1-4(2,3)5/h1-3H3 |
| SMIL | C[N+](C)(C)[O-] |
| IUPAC navn | N,N-dimethylmethanamine oxide |
| Molekylvægt (g/mol) | 75.11 |
| Formel vægt | 75.0684 |