missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylglycine methyl ester, 98%, Thermo Scientific Chemicals

Catalog Number p-7045764
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7148-06-3
MFCD00059303
This item is not returnable. View return policy
7148-06-3
MFCD00059303

N,N-Dimethylglycine methyl ester is used as a biochemical for proteomics research. It is also used as chemical and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylglycine methyl ester is used as a biochemical for proteomics research. It is also used as chemical and pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
It is sensitive to air and hygroscopic in nature. Protect from humidity and water. Store under dry inert gas. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Boiling Point 134°C to 135°C
Molecular Formula C5H11NO2
Linear Formula (CH3)2NCH2CO2CH3
UN Number UN3272
Beilstein 1744788
Synonym n,n-dimethylglycine methyl ester, methyl 2-dimethylamino acetate, glycine, n,n-dimethyl-, methyl ester, methyl dimethylamino acetate, methyl n,n-dimethylglycinate, acmc-2097kq, dimethylglycine methyl ester, ch3 2nch2cooch3, dimethylaminoessigsauremethylester
Refractive Index 1.412
Solubility Information Soluble in water.
InChI Key LRZFEBJUJIQVDQ-UHFFFAOYSA-N
SMILES CN(C)CC(=O)OC
IUPAC Name methyl 2-(dimethylamino)acetate
Molecular Weight (g/mol) 117.15
PubChem CID 81569
Formula Weight 117.15
Percent Purity 98%
Density 0.95
Quantity 5 g
Flash Point 38°C (100°F)
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material N,N-Dimethylglycine methyl ester
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'