Få mere at vide
N,N-Dimethylglycine hydrochloride, Sigma-Aldrich™
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N,N-Dimethylglycine hydrochloride |
| Smeltepunkt | 189°C to 194°C (External MSDS) |
| CAS | 2491-06-7 |
| Molekylær formel | (CH3)2NCH2COOH · HCl |
| Synonym | N,N-Dimethylglycine hydrochloride, N,N-Dimethylaminoacetic acid hydrochloride, N,N-Dimethylglycine hydrochloride |
| Molekylvægt (g/mol) | 139.58 |
| Mængde | 10 g |