Få mere at vide
N,N-Dimethyldecylamine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N,N-Dimethyldecylamine |
| CAS | 1120-24-7 |
| Molekylær formel | C12H27N |
| Mængde | 25 mL |
| Synonym | N,N-Dimethyl-1-decanamine,, N,N-Dimethyl-decylamine, 1-Dimethylaminodecane, ADMA 10, Armeen DM 10D, Decyldimethylamine, Dimethyldecylamine, Farmin DM 1098, Farmin DM 10P, N,N-Dimethyl-N-decylamine, N,N-Dimethyldecylamine, N-Decyl-N,N-dimethylamine, NSC 24529 |
| InChI formel | InChI=1S/C12H27N/c1-4-5-6-7-8-9-10-11-12-13(2)3/h4-12H2,1-3H3 |
| SMIL | CN(CCCCCCCCCC)C |
| IUPAC navn | N,N-dimethyldecan-1-amine |
| Molekylvægt (g/mol) | 185.35 |
| Formel vægt | 185.2143 |