Få mere at vide
N,N-Dimethylaniline-d6, TRC
Artikelnummer. 30375428
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30375428 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375428 Leverandør Toronto Research Chemicals Leverandørnr. TRCD464102250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethylaniline-d6
CAS 4019-61-8
Molekylær formel C8 D6 H5 N
Mængde 250 mg
Synonym Benzenamine, N,N-di(methyl-d3)- (9CI, ACI), Aniline, N,N-di(methyl-d3)- (7CI, 8CI), N,N-Di(methyl-d3)benzenamine (ACI), N,N-Bis(trideuteriomethyl)aniline, N,N-Dimethyl-d6-aniline, N,N-Dimethylaniline-d6, N,N-Dimethylaniline-d6, N,N-Dimethylaniline-D6 (N,N-dimethyl-D6), N,N-Di(methyl-d3)aniline
InChI formel InChI=1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])N(c1ccccc1)C([2H])([2H])[2H]
IUPAC navn N,N-bis(trideuteriomethyl)aniline
Molekylvægt (g/mol) 127.217
Formel vægt 127.127